Assay Details
Target Gene Details
Entrez Gene ID: | 79834 |
Gene Name: | pseudopodium enriched atypical kinase 1 |
Gene Aliases: |
SGK269 |
Location: |
Chr.15:77100829-77420104 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEAK1 | NM_024776.3 | 8 | 11405 | NP_079052.2 |
| XM_005254670.2 | 11 | 11709 | XP_005254727.1 | |
| XM_005254671.2 | 10 | 11606 | XP_005254728.1 | |
| XM_005254673.2 | 8 | 11340 | XP_005254730.1 | |
| XM_005254674.2 | 9 | 11553 | XP_005254731.1 | |
| XM_005254675.4 | 5 | 11059 | XP_005254732.1 | |
| XM_011522033.2 | 12 | 11765 | XP_011520335.1 | |
| XM_011522034.2 | 13 | 11873 | XP_011520336.1 | |
| XM_011522035.2 | 15 | 12024 | XP_011520337.1 | |
| XM_011522036.2 | 14 | 11928 | XP_011520338.1 | |
| XM_011522037.2 | 14 | 11969 | XP_011520339.1 | |
| XM_011522038.2 | 13 | 11820 | XP_011520340.1 | |
| XM_011522039.2 | 14 | 11916 | XP_011520341.1 | |
| XM_011522040.1 | XP_011520342.1 | |||
| XM_011522042.2 | 4 | 8142 | XP_011520344.1 | |
| AB082533.1 | 4 | 7884 | BAC02711.1 | |
| AK025943.1 | 1 | 1339 | ||
| AK124548.1 | 1 | 3241 | ||
| AW069191.1 | 1 | 430 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map