Assay Details
Target Gene Details
Entrez Gene ID: | 123624 |
Gene Name: | ATP/GTP binding protein like 1 |
Gene Aliases: |
CCP4, FECD8 |
Location: |
Chr.15:86079635-87031476 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL1 | XM_011521226.2 | XP_011519528.1 | ||
| XM_011521227.2 | XP_011519529.1 | |||
| XM_017021918.1 | XP_016877407.1 | |||
| XM_017021919.1 | XP_016877408.1 | |||
| XM_017021920.1 | XP_016877409.1 | |||
| DY655510.1 |
Target Gene Details
Entrez Gene ID: | 727915 |
Gene Name: | AGBL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:86296833-86316968 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL1-AS1 | NR_046012.1 | |||
| CD359326.1 | ||||
| DB524499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833080 | Chr.15:86175034 - 86343707 on Build GRCh38 | Gain |
|
| nsv519797 | Chr.15:86084875 - 87238541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map