Assay Details
Target Gene Details
Entrez Gene ID: | 58489 |
Gene Name: | abhydrolase domain containing 17C |
Gene Aliases: |
FAM108C1 |
Location: |
Chr.15:80695311-80755621 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD17C | NM_021214.1 | NP_067037.1 | ||
| AK298606.1 | ||||
| AL390079.1 | CAB98203.1 | |||
| BC059401.1 | ||||
| BC115003.1 | AAI15004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471685 | Chr.15:80530860 - 80711048 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map