Assay Details
Target Gene Details
Entrez Gene ID: | 10509 |
Gene Name: | semaphorin 4B |
Gene Aliases: |
SEMAC, SemC |
Location: |
Chr.15:90184920-90229660 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA4B | NM_001324029.1 | NP_001310958.1 | ||
| NM_001324030.1 | NP_001310959.1 | |||
| NM_001324031.1 | NP_001310960.1 | |||
| NM_001324032.1 | NP_001310961.1 | |||
| NM_001324033.1 | NP_001310962.1 | |||
| NM_001324034.1 | NP_001310963.1 | |||
| NM_020210.3 | NP_064595.2 | |||
| NM_198925.2 | NP_945119.1 | |||
| AB051532.1 | ||||
| AK026108.1 | ||||
| AK289991.1 | ||||
| AY358392.1 | AAQ88758.1 | |||
| AY445887.1 | AAR15707.1 | |||
| BC110853.1 | AAI10854.1 | |||
| BC126438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637214 | Chr.15:90184741 - 90235676 on Build GRCh38 | Gain |
|
| nsv833087 | Chr.15:90208675 - 90425725 on Build GRCh38 | Loss |
|
| nsv1052652 | Chr.15:90101186 - 90298961 on Build GRCh38 | Gain |
|
| dgv112e55 | Chr.15:90079272 - 90249274 on Build GRCh38 | Gain |
|
| esv2760054 | Chr.15:90022724 - 90266401 on Build GRCh38 | Gain |
|
| nsv817707 | Chr.15:90104624 - 90241698 on Build GRCh38 | Gain |
|
| nsv442384 | Chr.15:90089378 - 90235161 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150400707,rs76356360] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map