Assay Details
Target Gene Details
Entrez Gene ID: | 101929743 |
Gene Name: | long intergenic non-protein coding RNA 1586 |
Gene Aliases: |
- |
Location: |
Chr.15:88585571-88605110 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01586 | NR_120369.1 | |||
| NR_120370.1 | ||||
| AK054749.1 | ||||
| BX488539.1 |

Genomic Map