Assay Details
Target Gene Details
Entrez Gene ID: | 90527 |
Gene Name: | dual oxidase maturation factor 1 |
Gene Aliases: |
NIP, NUMBIP, mol |
Location: |
Chr.15:45117366-45129938 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUOXA1 | NM_001276264.1 | NP_001263193.1 | ||
| NM_001276265.1 | NP_001263194.1 | |||
| NM_001276267.1 | 9 | 1925 | NP_001263196.1 | |
| NM_001276268.1 | 6 | 1585 | NP_001263197.1 | |
| NM_144565.3 | NP_653166.2 | |||
| XM_006720743.3 | 9 | 2255 | XP_006720806.1 | |
| XM_006720745.1 | 9 | 2230 | XP_006720808.1 | |
| XM_006720746.1 | 9 | 2259 | XP_006720809.1 | |
| XM_006720747.3 | 8 | 2167 | XP_006720810.1 | |
| XM_006720749.1 | 9 | 2025 | XP_006720812.1 | |
| XM_006720751.3 | 7 | 1748 | XP_006720814.1 | |
| XM_006720752.3 | 7 | 1757 | XP_006720815.2 | |
| XM_006720754.1 | 8 | 1890 | XP_006720817.1 | |
| XM_006720755.1 | 8 | 1866 | XP_006720818.1 | |
| XM_011522181.2 | 9 | 2283 | XP_011520483.1 | |
| XM_011522182.1 | 8 | 1998 | XP_011520484.1 | |
| XM_011522183.2 | 10 | 2113 | XP_011520485.1 | |
| AK056896.1 | 6 | 1585 | BAB71304.1 | |
| AK292771.1 | ||||
| BC029819.1 | AAH29819.1 | |||
| EU927396.1 | ||||
| EU927397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map