Assay Details
Target Gene Details
Entrez Gene ID: | 1723 |
Gene Name: | dihydroorotate dehydrogenase (quinone) |
Gene Aliases: |
DHOdehase, POADS, URA1 |
Location: |
Chr.16:72008744-72027659 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHODH | NM_001361.4 | NP_001352.2 | ||
| XM_005255827.3 | XP_005255884.1 | |||
| XM_017022990.1 | XP_016878479.1 | |||
| AK292293.1 | ||||
| AU130529.1 | ||||
| BC065245.1 | AAH65245.1 | |||
| BI253747.1 | ||||
| M94065.1 | AAA50163.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055854 | Chr.16:71888241 - 72048987 on Build GRCh38 | Loss |
|
| esv3582296 | Chr.16:71965845 - 72027933 on Build GRCh38 | Loss |
|
| nsv428329 | Chr.16:71942441 - 72240448 on Build GRCh38 | Gain |
|
| esv2714683 | Chr.16:71950823 - 72151945 on Build GRCh38 | Deletion |
|
| nsv833273 | Chr.16:71937902 - 72118955 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map