Assay Details
Target Gene Details
Entrez Gene ID: | 4094 |
Gene Name: | MAF bZIP transcription factor |
Gene Aliases: |
AYGRP, CCA4, CTRCT21, c-MAF |
Location: |
Chr.16:79202624-79600725 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAF |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639241 | Chr.16:79219778 - 79258687 on Build GRCh38 | Loss |
|
| esv2662766 | Chr.16:79226224 - 79245902 on Build GRCh38 | Deletion |
|
| nsv573287 | Chr.16:79169293 - 79234387 on Build GRCh38 | Loss |
|
| esv3553726 | Chr.16:79226104 - 79248315 on Build GRCh38 | Deletion |
|
| esv3582366 | Chr.16:79226608 - 79241358 on Build GRCh38 | Loss |
|
| esv2762366 | Chr.16:79226621 - 79241452 on Build GRCh38 | Loss |
|
| nsv1063199 | Chr.16:79224428 - 79248434 on Build GRCh38 | Loss |
|
| esv2760426 | Chr.16:79218610 - 79245768 on Build GRCh38 | Gain |
|
| esv3639243 | Chr.16:79224245 - 79245954 on Build GRCh38 | Gain |
|
| nsv573285 | Chr.16:79124285 - 80093429 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74036773] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map