Assay Details
Target Gene Details
Entrez Gene ID: | 102723510 |
Gene Name: | LCMT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.16:25100564-25111555 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCMT1-AS1 | NR_120306.1 | |||
| AL600125.1 | ||||
| BM790349.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2827n100 | Chr.16:25051972 - 25126693 on Build GRCh38 | Gain |
|
| esv3441 | Chr.16:25054733 - 25126265 on Build GRCh38 | Loss |
|
| esv2758638 | Chr.16:24932074 - 25175657 on Build GRCh38 | Gain+Loss |
|
| esv3638267 | Chr.16:25097954 - 25119835 on Build GRCh38 | Gain |
|
| esv2714121 | Chr.16:25054771 - 25126057 on Build GRCh38 | Deletion |
|
| dgv48e19 | Chr.16:25054716 - 25128306 on Build GRCh38 | Loss |
|
| esv2660591 | Chr.16:25054775 - 25125976 on Build GRCh38 | Deletion |
|
| esv34994 | Chr.16:25057265 - 25125931 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112106314] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map