Assay Details
Target Gene Details
Entrez Gene ID: | 6530 |
Gene Name: | solute carrier family 6 member 2 |
Gene Aliases: |
NAT1, NET, NET1, SLC6A5 |
Location: |
Chr.16:55655604-55706192 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A2 | NM_001043.3 | NP_001034.1 | ||
| NM_001172501.1 | NP_001165972.1 | |||
| NM_001172504.1 | NP_001165975.1 | |||
| XM_006721263.2 | XP_006721326.1 | |||
| XM_011523295.1 | XP_011521597.1 | |||
| XM_011523296.1 | XP_011521598.1 | |||
| XM_011523297.2 | XP_011521599.1 | |||
| XM_011523298.1 | XP_011521600.1 | |||
| XM_011523300.2 | 1 | 654 | XP_011521602.1 | |
| AK312793.1 | ||||
| AU132072.1 | ||||
| M65105.1 | AAA59943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571298 | Chr.16:55605227 - 55802076 on Build GRCh38 | Gain |
|
| esv2758649 | Chr.16:55635484 - 55949678 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map