Assay Details
Target Gene Details
Entrez Gene ID: | 2653 |
Gene Name: | glycine cleavage system protein H |
Gene Aliases: |
GCE, NKH |
Location: |
Chr.16:81081947-81096375 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCSH | NM_004483.4 | NP_004474.2 | ||
| NR_033249.1 | ||||
| XM_017023136.1 | XP_016878625.1 | |||
| AK311978.1 | ||||
| AV696698.1 | ||||
| BF798008.1 | ||||
| BG708388.1 | ||||
| BT009827.1 | AAP88829.1 | |||
| CN283805.1 | ||||
| CN283809.1 | ||||
| CR457072.1 | CAG33353.1 | |||
| M69175.1 | AAA36011.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751615 | Chr.16:81059494 - 81273994 on Build GRCh38 | Gain |
|
| nsv1062724 | Chr.16:81018707 - 81103402 on Build GRCh38 | Gain |
|
| esv2422360 | Chr.16:81032428 - 81147122 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75633863] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map