Assay Details
Target Gene Details
Entrez Gene ID: | 283932 |
Gene Name: | FBXL19 antisense RNA 1 (head to head) |
Gene Aliases: |
NCRNA00095 |
Location: |
Chr.16:30919319-30923269 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL19-AS1 | NR_024348.1 | 1 | 470 | |
| AK092263.1 | 1 | 470 | BAC03841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss |
|
| nsv571820 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
| nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion |
|
| nsv833189 | Chr.16:30808387 - 30949474 on Build GRCh38 | Loss |
|

Genomic Map