Assay Details
Target Gene Details
Entrez Gene ID: | 100128788 |
Gene Name: | SRRM2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.16:2737076-2752600 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRM2-AS1 | NR_027274.1 | |||
| NR_027275.1 | ||||
| AK056063.1 | ||||
| AK098626.1 | ||||
| AK125806.1 | 1 | 2286 | BAC86301.1 | |
| DB448439.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
| esv3637636 | Chr.16:2737650 - 2739977 on Build GRCh38 | Loss |
|
| nsv471073 | Chr.16:2687070 - 2773569 on Build GRCh38 | Loss |
|
| nsv833123 | Chr.16:2561247 - 2764988 on Build GRCh38 | Gain+Loss |
|

Genomic Map