Assay Details
Target Gene Details
Entrez Gene ID: | 146227 |
Gene Name: | brain expressed, associated with NEDD4, 1 |
Gene Aliases: |
BEAN, SCA31 |
Location: |
Chr.16:66427297-66495288 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BEAN1 | NM_001136106.4 | NP_001129578.1 | ||
| NM_001178020.2 | NP_001171491.1 | |||
| NM_001197224.3 | NP_001184153.1 | |||
| NM_001197225.3 | NP_001184154.1 | |||
| XM_011522883.1 | XP_011521185.1 | |||
| XM_011522884.1 | XP_011521186.1 | |||
| XM_011522887.2 | XP_011521189.1 | |||
| XM_011522888.1 | XP_011521190.1 | |||
| XM_011522889.1 | XP_011521191.1 | |||
| XM_011522890.1 | XP_011521192.1 | |||
| XM_011522891.1 | XP_011521193.1 | |||
| XM_011522892.2 | XP_011521194.1 | |||
| XM_011522895.1 | XP_011521197.1 | |||
| AB472390.1 | ||||
| AB472391.1 | ||||
| AB472392.1 | ||||
| AB472394.1 | ||||
| AB472396.1 | ||||
| AB472399.1 | ||||
| AK056142.1 | ||||
| BC107477.1 | AAI07478.1 |
Target Gene Details
Entrez Gene ID: | 101927726 |
Gene Name: | BEAN1 antisense RNA 1 |
Gene Aliases: |
CTD-2258A20.5 |
Location: |
Chr.16:66469796-66476402 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BEAN1-AS1 | NR_109960.1 | |||
| DA850942.1 | ||||
| DA859002.1 | ||||
| DA862085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833264 | Chr.16:66298692 - 66471069 on Build GRCh38 | Gain |
|

Genomic Map