Assay Details
Target Gene Details
Entrez Gene ID: | 11269 |
Gene Name: | DEAD-box helicase 19B |
Gene Aliases: |
DBP5, DDX19, RNAh |
Location: |
Chr.16:70289767-70335213 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX19B | NM_001257174.1 | NP_001244103.1 | ||
| AF353720.1 | AAK40102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061286 | Chr.16:70219906 - 70297504 on Build GRCh38 | Gain |
|
| nsv833271 | Chr.16:70151791 - 70319431 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74578812] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map