Assay Details
Target Gene Details
Entrez Gene ID: | 23450 |
Gene Name: | splicing factor 3b subunit 3 |
Gene Aliases: |
RSE1, SAP130, SF3b130, STAF130 |
Location: |
Chr.16:70523788-70577668 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B3 | NM_012426.4 | NP_036558.3 | ||
| AJ001443.1 | CAB56791.1 | |||
| AK291768.1 | ||||
| BC009780.1 | AAH09780.1 | |||
| BC068974.1 | AAH68974.1 | |||
| D87686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833272 | Chr.16:70335866 - 70535820 on Build GRCh38 | Loss |
|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
| esv3892883 | Chr.16:70513090 - 70660097 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map