Assay Details
Target Gene Details
Entrez Gene ID: | 9289 |
Gene Name: | adhesion G protein-coupled receptor G1 |
Gene Aliases: |
BFPP, BPPR, GPR56, TM7LN4, TM7XN1 |
Location: |
Chr.16:57619535-57665039 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065634 | Chr.16:57638274 - 57670341 on Build GRCh38 | Gain |
|
| nsv833246 | Chr.16:57577216 - 57747868 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map