Assay Details
Target Gene Details
Entrez Gene ID: | 64711 |
Gene Name: | heparan sulfate-glucosamine 3-sulfotransferase 6 |
Gene Aliases: |
3-OST-6, HS3ST5 |
Location: |
Chr.16:1911463-1921428 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HS3ST6 | NM_001009606.3 | NP_001009606.3 | ||
| XM_011522608.2 | XP_011520910.1 | |||
| DY654407.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| nsv952901 | Chr.16:1888800 - 2135899 on Build GRCh38 | Deletion |
|
| nsv519366 | Chr.16:1851210 - 1961125 on Build GRCh38 | Loss |
|
| nsv833120 | Chr.16:1893102 - 1980660 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116116044] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map