Assay Details
Target Gene Details
Entrez Gene ID: | 51728 |
Gene Name: | RNA polymerase III subunit K |
Gene Aliases: |
C11, C11-RNP3, My010, RPC10, RPC11, RPC12.5 |
Location: |
Chr.16:46979-53632 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3K | NM_016310.4 | NP_057394.3 | ||
| AF051316.1 | AAF18268.1 | |||
| AF060223.1 | AAG43123.1 | |||
| BC011932.2 | AAH11932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518291 | Chr.16:47354 - 254515 on Build GRCh38 | Loss |
|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| nsv1044732 | Chr.16:35815 - 60707 on Build GRCh38 | Loss |
|
| esv3637535 | Chr.16:47684 - 48970 on Build GRCh38 | Loss |
|
| dgv2688n100 | Chr.16:10765 - 80854 on Build GRCh38 | Gain |
|
| esv2760065 | Chr.16:31052 - 143601 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74003634] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map