Assay Details
Target Gene Details
Entrez Gene ID: | 55308 |
Gene Name: | DEAD-box helicase 19A |
Gene Aliases: |
DDX19-DDX19L, DDX19L |
Location: |
Chr.16:70346829-70373383 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX19A | NM_001320522.1 | NP_001307451.1 | ||
| NM_001320525.1 | NP_001307454.1 | |||
| NM_001320526.1 | NP_001307455.1 | |||
| NM_001320527.1 | NP_001307456.1 | |||
| NM_018332.4 | NP_060802.1 | |||
| NR_135296.1 | ||||
| AK001988.1 | BAA92022.1 | |||
| AK223262.1 | BAD96982.1 | |||
| AK299539.1 | ||||
| AK310604.1 | ||||
| AK310804.1 | ||||
| AL832970.1 | CAH10622.1 | |||
| BC005162.2 | AAH05162.1 | |||
| BC006544.2 | AAH06544.1 | |||
| BC137496.1 | ||||
| BC137497.1 | ||||
| CR749227.1 | CAH18083.1 | |||
| DB074731.1 | ||||
| DC375258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833272 | Chr.16:70335866 - 70535820 on Build GRCh38 | Loss |
|
| esv2714670 | Chr.16:70331662 - 70370229 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map