Assay Details
Target Gene Details
Entrez Gene ID: | 11269 |
Gene Name: | DEAD-box helicase 19B |
Gene Aliases: |
DBP5, DDX19, RNAh |
Location: |
Chr.16:70289767-70335213 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX19B | NM_001257172.1 | NP_001244101.1 | ||
| NM_001257173.1 | NP_001244102.1 | |||
| NM_001257174.1 | NP_001244103.1 | |||
| XM_011522832.2 | XP_011521134.1 | |||
| XM_011522833.2 | XP_011521135.1 | |||
| XM_011522834.2 | XP_011521136.1 | |||
| XM_017022890.1 | XP_016878379.1 | |||
| AF353720.1 | AAK40102.1 | |||
| AK301938.1 | ||||
| AK302107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833271 | Chr.16:70151791 - 70319431 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map