Assay Details
Target Gene Details
Entrez Gene ID: | 763 |
Gene Name: | carbonic anhydrase 5A |
Gene Aliases: |
CA5, CA5AD, CAV, CAVA, GS1-21A4.1 |
Location: |
Chr.16:87888019-87936575 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA5A | NM_001739.1 | NP_001730.1 | ||
| XM_005256134.4 | XP_005256191.1 | |||
| XM_011523309.2 | XP_011521611.1 | |||
| XM_011523310.2 | XP_011521612.1 | |||
| XM_017023646.1 | XP_016879135.1 | |||
| BC137405.1 | ||||
| BC137411.1 | ||||
| L19297.1 | AAA02890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059786 | Chr.16:87811419 - 88019505 on Build GRCh38 | Gain |
|
| nsv9471 | Chr.16:87881137 - 87941142 on Build GRCh38 | Gain |
|
| esv3892935 | Chr.16:87808912 - 88010269 on Build GRCh38 | Loss |
|
| esv2758668 | Chr.16:87885744 - 88080126 on Build GRCh38 | Loss |
|
| nsv1066924 | Chr.16:87911850 - 87998507 on Build GRCh38 | Loss |
|
| nsv1062946 | Chr.16:87635801 - 88019505 on Build GRCh38 | Gain |
|
| dgv36n68 | Chr.16:87801736 - 87983550 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map