Assay Details
Target Gene Details
Entrez Gene ID: | 55890 |
Gene Name: | G protein-coupled receptor class C group 5 member C |
Gene Aliases: |
RAIG-3, RAIG3 |
Location: |
Chr.17:74431341-74451658 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRC5C | NM_018653.3 | NP_061123.3 | ||
| NM_022036.2 | NP_071319.2 | |||
| XM_005257512.1 | XP_005257569.1 | |||
| XM_005257514.1 | XP_005257571.1 | |||
| XM_017024855.1 | XP_016880344.1 | |||
| XM_017024856.1 | XP_016880345.1 | |||
| AB451470.1 | ||||
| AF207989.1 | AAF72870.1 | |||
| AJ276102.1 | CAC00633.1 | |||
| AK000249.1 | BAA91030.1 | |||
| AK075397.1 | BAC11595.1 | |||
| AK131210.1 | ||||
| AK222909.1 | ||||
| BC004925.1 | AAH04925.1 | |||
| BC016860.1 | AAH16860.2 | |||
| BC110848.1 | AAI10849.1 | |||
| BC125080.1 | ||||
| BC125081.1 | ||||
| BT007435.1 | AAP36103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12e196 | Chr.17:73675043 - 74565539 on Build GRCh38 | Duplication |
|
| esv3641237 | Chr.17:74241755 - 74451924 on Build GRCh38 | Gain |
|
| esv2761978 | Chr.17:73835799 - 74684915 on Build GRCh38 | Gain |
|
| esv3641230 | Chr.17:73831787 - 74694861 on Build GRCh38 | Gain |
|
| dgv3282n100 | Chr.17:73814343 - 74719597 on Build GRCh38 | Gain |
|
| esv33995 | Chr.17:74261053 - 74732093 on Build GRCh38 | Loss |
|
| dgv372n27 | Chr.17:73826016 - 74685802 on Build GRCh38 | Gain |
|
| esv2751770 | Chr.17:73837862 - 74716223 on Build GRCh38 | Gain |
|
| nsv952366 | Chr.17:74429862 - 74456261 on Build GRCh38 | Deletion |
|
| esv3573820 | Chr.17:73817932 - 74694378 on Build GRCh38 | Gain |
|
| dgv5774n54 | Chr.17:74428381 - 74673021 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map