Assay Details
Target Gene Details
Entrez Gene ID: | 100128977 |
Gene Name: | MAPT antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:45843356-45895513 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPT-AS1 | NR_024559.1 | |||
| AW770488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518110 | Chr.17:45847931 - 45849423 on Build GRCh38 | Loss |
|
| esv3554471 | Chr.17:45847635 - 45852134 on Build GRCh38 | Deletion |
|
| esv2762433 | Chr.17:45496768 - 46710828 on Build GRCh38 | Gain+Loss |
|
| esv3640674 | Chr.17:45847682 - 45851860 on Build GRCh38 | Loss |
|
| esv2669509 | Chr.17:45847702 - 45851880 on Build GRCh38 | Deletion |
|
| nsv833460 | Chr.17:45798585 - 45893131 on Build GRCh38 | Loss |
|
| nsv575141 | Chr.17:45846971 - 45850758 on Build GRCh38 | Loss |
|

Genomic Map