Assay Details
Target Gene Details
Entrez Gene ID: | 100287042 |
Gene Name: | uncharacterized LOC100287042 |
Gene Aliases: |
- |
Location: |
Chr.17:75271299-75273895 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100287042 | NR_036520.1 | 1 | 881 | |
| AK296245.1 | 1 | 811 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
|
| nsv1066312 | Chr.17:75260266 - 75323694 on Build GRCh38 | Gain |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|

Genomic Map