Assay Details
Target Gene Details
Entrez Gene ID: | 642843 |
Gene Name: | cleavage and polyadenylation specific factor 4 like |
Gene Aliases: |
- |
Location: |
Chr.17:73242343-73263778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPSF4L | NM_001129885.1 | NP_001123357.1 | ||
| XM_011525115.2 | XP_011523417.1 | |||
| BC157870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2138 | Chr.17:73234909 - 73269410 on Build GRCh38 | Insertion |
|
| nsv833534 | Chr.17:73155642 - 73349355 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map