Assay Details
Target Gene Details
Entrez Gene ID: | 4804 |
Gene Name: | nerve growth factor receptor |
Gene Aliases: |
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR |
Location: |
Chr.17:49495293-49515020 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGFR | NM_002507.3 | NP_002498.1 | ||
| AK303278.1 | ||||
| AK313654.1 | ||||
| BC050309.1 | AAH50309.1 | |||
| DA115173.1 | ||||
| M14764.1 | AAB59544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833473 | Chr.17:49383665 - 49545926 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72837241] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map