Assay Details
Target Gene Details
Entrez Gene ID: | 124935 |
Gene Name: | solute carrier family 43 member 2 |
Gene Aliases: |
LAT4 |
Location: |
Chr.17:1569254-1630014 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC43A2 | NM_001284498.1 | 15 | 5843 | NP_001271427.1 |
| NM_001284499.1 | 10 | 5284 | NP_001271428.1 | |
| NM_001321364.1 | 15 | 5641 | NP_001308293.1 | |
| NM_001321365.1 | 15 | 5810 | NP_001308294.1 | |
| NM_152346.2 | 14 | 5629 | NP_689559.1 | |
| AK056769.1 | 1 | 721 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|
| nsv1945 | Chr.17:1560608 - 1589052 on Build GRCh38 | Insertion |
|
| esv1004030 | Chr.17:1566596 - 1574887 on Build GRCh38 | Deletion |
|
| nsv457635 | Chr.17:1554545 - 1638896 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112833601] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map