Assay Details
Target Gene Details
Entrez Gene ID: | 100288866 |
Gene Name: | uncharacterized LOC100288866 |
Gene Aliases: |
- |
Location: |
Chr.17:49505657-49574064 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100288866 | NR_103773.1 | |||
| AK303671.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833473 | Chr.17:49383665 - 49545926 on Build GRCh38 | Gain+Loss |
|

Genomic Map