Assay Details
Target Gene Details
Entrez Gene ID: | 26207 |
Gene Name: | phosphatidylinositol transfer protein, cytoplasmic 1 |
Gene Aliases: |
M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1 |
Location: |
Chr.17:67377281-67697263 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITPNC1 | NM_012417.3 | NP_036549.2 | ||
| NM_181671.2 | NP_858057.1 | |||
| XM_005257216.2 | XP_005257273.1 | |||
| XM_017024444.1 | XP_016879933.1 | |||
| AF171102.1 | AAF06148.1 | |||
| AK290838.1 | ||||
| BC007905.2 | AAH07905.1 | |||
| BC067095.1 | AAH67095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23559 | Chr.17:67392080 - 67405562 on Build GRCh38 | Loss |
|
| nsv471703 | Chr.17:67393510 - 67554789 on Build GRCh38 | Gain |
|
| dgv5751n54 | Chr.17:67393189 - 67405433 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map