Assay Details
Target Gene Details
Entrez Gene ID: | 100130950 |
Gene Name: | uncharacterized LOC100130950 |
Gene Aliases: |
- |
Location: |
Chr.17:5192084-5235636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130950 | NR_034082.1 | |||
| AK021878.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| dgv3089n100 | Chr.17:5187013 - 5327634 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73333445] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map