Assay Details
Target Gene Details
Entrez Gene ID: | 26151 |
Gene Name: | N-acetyltransferase 9 (putative) |
Gene Aliases: |
EBSP, hNATL |
Location: |
Chr.17:74770529-74776425 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT9 | NM_001305077.1 | NP_001292006.1 | ||
| NM_001305078.1 | NP_001292007.1 | |||
| NM_001305079.1 | NP_001292008.1 | |||
| NM_001305080.1 | NP_001292009.1 | |||
| NM_001305081.1 | NP_001292010.1 | |||
| NM_001305082.1 | NP_001292011.1 | |||
| NM_001305083.1 | NP_001292012.1 | |||
| NM_001305084.1 | NP_001292013.1 | |||
| NM_001305085.1 | NP_001292014.1 | |||
| NM_001305086.1 | NP_001292015.1 | |||
| NM_001305087.1 | NP_001292016.1 | |||
| NM_001305088.1 | NP_001292017.1 | |||
| NM_015654.4 | NP_056469.2 | |||
| NR_130953.1 | ||||
| NR_130954.1 | ||||
| HY222854.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map