Assay Details
Target Gene Details
Entrez Gene ID: | 2081 |
Gene Name: | endoplasmic reticulum to nucleus signaling 1 |
Gene Aliases: |
IRE1, IRE1P, IRE1a, hIRE1p |
Location: |
Chr.17:64039142-64130819 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERN1 | XM_017024347.1 | 22 | 6002 | XP_016879836.1 |
| XM_017024348.1 | 21 | 5900 | XP_016879837.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833511 | Chr.17:63865995 - 64064305 on Build GRCh38 | Loss |
|
| nsv483108 | Chr.17:64041483 - 64196113 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map