Assay Details
Target Gene Details
Entrez Gene ID: | 4137 |
Gene Name: | microtubule associated protein tau |
Gene Aliases: |
DDPAC, FTDP-17, MAPTL, MSTD, MTBT1, MTBT2, PPND, PPP1R103, TAU |
Location: |
Chr.17:45894382-46028334 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762433 | Chr.17:45496768 - 46710828 on Build GRCh38 | Gain+Loss |
|
| dgv3178n100 | Chr.17:45877004 - 46034461 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114728005] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map