Assay Details
Target Gene Details
Entrez Gene ID: | 339221 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 7 |
Gene Aliases: |
ALK-SMase, E-NPP 7, NPP-7, NPP7 |
Location: |
Chr.17:79730919-79742926 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP7 | XM_011524737.1 | XP_011523039.1 |
More Information
Set Membership: |
|

Genomic Map