Assay Details
Target Gene Details
Entrez Gene ID: | 100507410 |
Gene Name: | C1QTNF1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:79019209-79027655 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QTNF1-AS1 | NR_040018.1 | |||
| NR_040019.1 | ||||
| DA938488.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470610 | Chr.17:78973800 - 79091723 on Build GRCh38 | Loss |
|
| nsv523650 | Chr.17:78933797 - 79165563 on Build GRCh38 | Loss |
|
| nsv1057193 | Chr.17:78921031 - 79481744 on Build GRCh38 | Gain |
|

Genomic Map