Assay Details
Target Gene Details
Entrez Gene ID: | 10209 |
Gene Name: | eukaryotic translation initiation factor 1 |
Gene Aliases: |
A121, EIF-1, EIF1A, ISO1, SUI1 |
Location: |
Chr.17:41688875-41691646 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1 | NM_005801.3 | NP_005792.1 | ||
| AF083441.1 | AAD52028.1 | |||
| AF100737.1 | AAD19900.1 | |||
| AJ012375.1 | CAD66615.1 | |||
| AK311209.1 | ||||
| AK311862.1 | ||||
| AL050005.1 | 1 | 537 | ||
| BC005118.1 | AAH05118.1 | |||
| BC008710.1 | AAH08710.1 | |||
| BT006645.1 | AAP35291.1 | |||
| BU165055.1 | ||||
| CR457051.1 | CAG33332.1 | |||
| L26247.1 | AAA60602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map