Assay Details
Target Gene Details
Entrez Gene ID: | 100306951 |
Gene Name: | PITPNA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:1516919-1518096 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITPNA-AS1 | NR_028514.1 | |||
| AA341969.1 | ||||
| DB053306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|

Genomic Map