Assay Details
Target Gene Details
Entrez Gene ID: | 440465 |
Gene Name: | BAIAP2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.17:81029133-81034719 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAIAP2-AS1 | NR_026857.1 | |||
| AK056555.1 | ||||
| AK124762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828128 | Chr.17:80659041 - 81736487 on Build GRCh38 | Loss |
|
| nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss |
|
| nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss |
|
| nsv576169 | Chr.17:81030816 - 81043967 on Build GRCh38 | Loss |
|
| dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117814967] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map