Assay Details
Target Gene Details
Entrez Gene ID: | 284021 |
Gene Name: | mast cell immunoglobulin like receptor 1 |
Gene Aliases: |
Allergin-1, C17orf60, MCA-32, MCA32 |
Location: |
Chr.17:64449022-64474009 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MILR1 | NM_001085423.1 | NP_001078892.1 | ||
| NM_001291316.1 | NP_001278245.1 | |||
| NM_001291317.1 | NP_001278246.1 | |||
| XM_017024486.1 | XP_016879975.1 | |||
| XM_017024487.1 | XP_016879976.1 | |||
| AB542950.1 | ||||
| AB542951.1 | ||||
| AB542952.1 | ||||
| BC053534.1 | AAH53534.1 | |||
| BC130406.1 | ||||
| BC136594.1 | ||||
| BC144272.1 | ||||
| BE296352.1 |
More Information
Set Membership: |
|

Genomic Map