Assay Details
Target Gene Details
Entrez Gene ID: | 101928251 |
Gene Name: | uncharacterized LOC101928251 |
Gene Aliases: |
- |
Location: |
Chr.17:73513683-73518770 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928251 | NR_135635.1 |
Target Gene Details
Entrez Gene ID: | 54549 |
Gene Name: | sidekick cell adhesion molecule 2 |
Gene Aliases: |
- |
Location: |
Chr.17:73334384-73644482 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDK2 | NM_001144952.1 | NP_001138424.1 | ||
| XM_011524914.2 | XP_011523216.1 | |||
| XM_011524915.2 | XP_011523217.1 | |||
| XM_011524916.2 | XP_011523218.1 |
More Information
Set Membership: |
|

Genomic Map