Assay Details
Target Gene Details
Entrez Gene ID: | 5026 |
Gene Name: | purinergic receptor P2X 5 |
Gene Aliases: |
LRH-1, P2X5, P2X5R |
Location: |
Chr.17:3673227-3696404 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX5 | NM_001204519.1 | NP_001191448.1 | ||
| NM_001204520.1 | NP_001191449.1 | |||
| NM_002561.3 | NP_002552.2 | |||
| NM_175080.2 | NP_778255.1 | |||
| AF016709.1 | AAC51931.1 | |||
| AF070573.1 | AAC28645.1 | |||
| AK092966.1 | ||||
| AK290889.1 | ||||
| AK293610.1 | ||||
| AK307959.1 | ||||
| BC039015.1 | AAH39015.1 | |||
| DQ851492.1 | ABI31773.1 | |||
| U49395.1 | AAB08576.1 | |||
| U49396.1 | AAB08577.1 |
Target Gene Details
Entrez Gene ID: | 100533970 |
Gene Name: | P2RX5-TAX1BP3 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.17:3662893-3696404 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX5-TAX1BP3 | NR_037928.1 | |||
| AB209623.1 | BAD92860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
|
| nsv574237 | Chr.17:3486822 - 3729542 on Build GRCh38 | Loss |
|
| nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain |
|
| nsv1948 | Chr.17:3674767 - 3719479 on Build GRCh38 | Deletion |
|
| nsv833342 | Chr.17:3626188 - 3806082 on Build GRCh38 | Loss |
|
| nsv952107 | Chr.17:3640507 - 3691806 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74966626] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map