Assay Details
Target Gene Details
Entrez Gene ID: | 339221 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 7 |
Gene Aliases: |
ALK-SMase, E-NPP 7, NPP-7, NPP7 |
Location: |
Chr.17:79730919-79742926 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP7 | NM_178543.4 | NP_848638.3 | ||
| XM_011524737.1 | XP_011523039.1 | |||
| AK126250.1 | BAC86504.1 | |||
| AK128662.1 | 3 | 1221 | ||
| AY230663.1 | AAP69661.1 | |||
| AY358622.1 | AAQ88985.1 | |||
| BC041453.1 | AAH41453.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952381 | Chr.17:79677839 - 79741901 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map