Assay Details
Target Gene Details
Entrez Gene ID: | 80206 |
Gene Name: | formin homology 2 domain containing 3 |
Gene Aliases: |
FHOS2, Formactin2 |
Location: |
Chr.18:36297696-36780220 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Gene Details
Entrez Gene ID: | 105372071 |
Gene Name: | uncharacterized LOC105372071 |
Gene Aliases: |
- |
Location: |
Chr.18:36660389-36663418 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105372071 | NR_132982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576651 | Chr.18:36017567 - 36959615 on Build GRCh38 | Loss |
|
| esv2758722 | Chr.18:36470072 - 36901904 on Build GRCh38 | Gain |
|
| nsv817789 | Chr.18:36536338 - 36856407 on Build GRCh38 | Gain |
|
| esv3642236 | Chr.18:36659865 - 36881208 on Build GRCh38 | Loss |
|
| esv3583016 | Chr.18:36337333 - 37122274 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map