Assay Details
Target Gene Details
Entrez Gene ID: | 4729 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit V2 |
Gene Aliases: |
CI-24k |
Location: |
Chr.18:9102623-9134345 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFV2 | NM_021074.4 | NP_066552.2 | ||
| XM_017025782.1 | XP_016881271.1 | |||
| AF147367.1 | ||||
| AK291865.1 | ||||
| AK312129.1 | ||||
| AW051350.1 | ||||
| BC001632.1 | AAH01632.1 | |||
| BC017487.1 | AAH17487.1 | |||
| BC071689.1 | AAH71689.1 | |||
| BG742948.1 | ||||
| CR456928.1 | CAG33209.1 | |||
| M22538.1 | AAA75390.1 |
Target Gene Details
Entrez Gene ID: | 101927275 |
Gene Name: | NDUFV2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:9121265-9136720 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFV2-AS1 | NR_110771.1 | |||
| NR_110772.1 | ||||
| BI520375.1 | ||||
| HY015092.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73390120] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map