Assay Details
Target Gene Details
Entrez Gene ID: | 91768 |
Gene Name: | Cdk5 and Abl enzyme substrate 1 |
Gene Aliases: |
CABL1, CABLES, HsT2563, IK3-1 |
Location: |
Chr.18:23134564-23260470 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABLES1 | NM_001100619.2 | NP_001094089.1 | ||
| NM_001256438.1 | NP_001243367.1 | |||
| NM_138375.2 | NP_612384.1 | |||
| NR_023359.1 | ||||
| AF348525.1 | AAL83906.1 | |||
| AK093243.1 | BAC04107.1 | |||
| AK296407.1 | ||||
| AK304880.1 | ||||
| BC004124.2 | AAH04124.2 | |||
| BC037218.1 | AAH37218.1 | |||
| EF028204.1 |
Target Gene Details
Entrez Gene ID: | 85019 |
Gene Name: | transmembrane protein 241 |
Gene Aliases: |
C18orf45, hVVT |
Location: |
Chr.18:23237252-23437969 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM241 | XM_017026043.1 | XP_016881532.1 | ||
| XM_017026046.1 | XP_016881535.1 | |||
| XM_017026047.1 | XP_016881536.1 | |||
| XM_017026048.1 | XP_016881537.1 | |||
| AK294916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758716 | Chr.18:23190468 - 23347650 on Build GRCh38 | Loss |
|
| esv2761994 | Chr.18:23241557 - 23257139 on Build GRCh38 | Gain |
|
| esv32657 | Chr.18:23203968 - 23353903 on Build GRCh38 | Loss |
|
| dgv5873n54 | Chr.18:23220751 - 23299468 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map