Assay Details
Target Gene Details
Entrez Gene ID: | 100507003 |
Gene Name: | golgi associated olfactory signaling regulator |
Gene Aliases: |
Goofy |
Location: |
Chr.19:49425399-49429064 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFY | NM_001195256.1 | 1 | 853 | NP_001182185.1 |
| XM_011526299.2 | 2 | 965 | XP_011524601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv52n68 | Chr.19:49286685 - 49456131 on Build GRCh38 | Loss |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map