Assay Details
Target Gene Details
Entrez Gene ID: | 79709 |
Gene Name: | collagen beta(1-O)galactosyltransferase 1 |
Gene Aliases: |
GLT25D1 |
Location: |
Chr.19:17555594-17583162 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COLGALT1 | NM_024656.3 | NP_078932.2 | ||
| XM_005260080.4 | XP_005260137.1 | |||
| XM_011528297.2 | XP_011526599.1 | |||
| XM_011528298.2 | XP_011526600.1 | |||
| XM_011528299.2 | XP_011526601.1 | |||
| AK074941.1 | BAC11307.1 | |||
| AK075541.1 | BAC11684.1 | |||
| BC071684.1 | AAH71684.1 | |||
| BC108308.1 | AAI08309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422429 | Chr.19:17529868 - 17579053 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138151046] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map