Assay Details
Target Gene Details
Entrez Gene ID: | 342 |
Gene Name: | apolipoprotein C1 pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.19:44926803-44931386 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOC1P1 | NR_028412.1 | 2 | 478 | |
| NR_028413.1 | 2 | 293 | ||
| NR_028414.1 | 3 | 170 | ||
| AV645786.1 | 2 | 331 | ||
| CB162526.1 | 2 | 255 | ||
| DA636997.1 | 3 | 169 | ||
| DA638554.1 | 2 | 293 | ||
| EF553526.1 | 2 | 293 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509747 | Chr.19:44877936 - 44943426 on Build GRCh38 | Insertion |
|

Genomic Map