Assay Details
Target Gene Details
Entrez Gene ID: | 147710 |
Gene Name: | immunoglobulin superfamily member 23 |
Gene Aliases: |
- |
Location: |
Chr.19:44612577-44636781 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF23 | NM_001205280.1 | NP_001192209.1 | ||
| XM_017026317.1 | XP_016881806.1 | |||
| XM_017026318.1 | XP_016881807.1 | |||
| XM_017026319.1 | XP_016881808.1 | |||
| XM_017026320.1 | XP_016881809.1 | |||
| XM_017026321.1 | XP_016881810.1 | |||
| XM_017026323.1 | XP_016881812.1 | |||
| XM_017026324.1 | XP_016881813.1 | |||
| XM_017026325.1 | XP_016881814.1 | |||
| BX112065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062812 | Chr.19:44538275 - 44711525 on Build GRCh38 | Gain |
|
| nsv483005 | Chr.19:44530751 - 44705087 on Build GRCh38 | Loss |
|

Genomic Map